Learn More
MSC HexArmor Safety Glasses, Photochromic Lenses, Anti-Fog, Scratch-Resistant, 1 ea
Catalog No. 5000793102
Encompass
These safety glasses feature anti-fog and scratch-resistant photochromic polycarbonate lenses, designed with a half-frame for durability and comfort. They provide comprehensive UV protection and meet strict safety standards, making them suitable for diverse work environments.
  • Sleek, modern wraparound lens and low-profile frame with soft touch components.
  • Extended lens for added protection and peripheral vision.
  • Dual-injection molded soft-grip sidearms for comfort and a non-slip fit.
  • Proven anti-fog and scratch-resistant coating.
  • Premium optical clarity (Optical Class 1).
  • Soft nose piece for added comfort and a non-slip fit.
  • Premium metal-free hinge design.
  • Provides 100% UV protection.
  • Adaptable frame design for durable comfort.
Catalog No. 50-007-93102 Supplier MSC Supplier No. 85943454
These safety glasses feature anti-fog and scratch-resistant photochromic polycarbonate lenses, designed with a half-frame for durability and comfort. They provide comprehensive UV protection and meet strict safety standards, making them suitable for diverse work environments.
  • Sleek, modern wraparound lens and low-profile frame with soft touch components.
  • Extended lens for added protection and peripheral vision.
  • Dual-injection molded soft-grip sidearms for comfort and a non-slip fit.
  • Proven anti-fog and scratch-resistant coating.
  • Premium optical clarity (Optical Class 1).
  • Soft nose piece for added comfort and a non-slip fit.
  • Premium metal-free hinge design.
  • Provides 100% UV protection.
  • Adaptable frame design for durable comfort.