Learn More
MSC HexArmor Safety Glasses, Polycarbonate, Blue Mirror and Gray, 1 ea
Catalog No. 5000788725
Encompass
These safety glasses feature scratch-resistant polycarbonate lenses with blue mirror and gray tints. They are designed with a full-framed, traditional style and non-adjustable temples, providing reliable eye protection.
  • Scratch resistant coating
  • Polycarbonate lens material
  • Blue mirror and gray lenses
  • Full-framed design
  • Non-adjustable temples
  • UV protection
  • Meets CE EN166, CSA Z94.3, and ANSI/ISEA Z87.1 (+) - 2020 standards
Catalog No. 50-007-88725 Supplier MSC Supplier No. 42879221
These safety glasses feature scratch-resistant polycarbonate lenses with blue mirror and gray tints. They are designed with a full-framed, traditional style and non-adjustable temples, providing reliable eye protection.
  • Scratch resistant coating
  • Polycarbonate lens material
  • Blue mirror and gray lenses
  • Full-framed design
  • Non-adjustable temples
  • UV protection
  • Meets CE EN166, CSA Z94.3, and ANSI/ISEA Z87.1 (+) - 2020 standards