Learn More
MSC HexArmor Safety Glasses, Scratch Resistant, Polycarbonate, Clear, Half-Framed, 1 ea
Catalog No. 5000674898
Encompass
HexArmor® Safety Glasses are designed to provide eye protection during everyday operations. Their scratch resistant coatings help maintain lens clarity, ensuring reliable performance.
  • Scratch resistant coatings prevent lens damage from minor debris
  • Wraparound glasses style for comprehensive coverage
  • Half-framed design with a wrap around frame
  • Clear polycarbonate lenses with UV protection
  • Universal size with +2 diopter lenses
  • Durable frame made from thermoplastic rubber and polycarbonate
  • Bayonet temple type for comfortable wear
  • Meets ANSI/ISEA Z87.1 (+) - 2020, CE EN166, and CSA Z94.3 standards
Catalog No. 50-006-74898 Supplier MSC Supplier No. 50732205
HexArmor® Safety Glasses are designed to provide eye protection during everyday operations. Their scratch resistant coatings help maintain lens clarity, ensuring reliable performance.
  • Scratch resistant coatings prevent lens damage from minor debris
  • Wraparound glasses style for comprehensive coverage
  • Half-framed design with a wrap around frame
  • Clear polycarbonate lenses with UV protection
  • Universal size with +2 diopter lenses
  • Durable frame made from thermoplastic rubber and polycarbonate
  • Bayonet temple type for comfortable wear
  • Meets ANSI/ISEA Z87.1 (+) - 2020, CE EN166, and CSA Z94.3 standards