Learn More
MSC HexArmor Safety Glasses, VS300, Progressive 2.0 Diopter, TruShield, 1 Pair
Catalog No. 5000796341
Encompass
The VS300 Safety Glasses provide full coverage protection with optical clarity. Designed for comfort and durability, these glasses feature advanced lens technology and permanent anti-fog and scratch-resistant coatings, ideal for laboratory and industrial environments.
  • Seven side arm inclination levels and soft ear pieces
  • Integrated brow guard and soft flex nose loops for comfort
  • Dual-action permanent anti-fog and scratch-resistant coatings
  • Provides 100% UV protection and metal-free construction
  • LTX® Lens Technology for premium optical clarity
  • Progressive +2.0 diopter for magnification
Catalog No. 50-007-96341 Supplier MSC Supplier No. 10000487
The VS300 Safety Glasses provide full coverage protection with optical clarity. Designed for comfort and durability, these glasses feature advanced lens technology and permanent anti-fog and scratch-resistant coatings, ideal for laboratory and industrial environments.
  • Seven side arm inclination levels and soft ear pieces
  • Integrated brow guard and soft flex nose loops for comfort
  • Dual-action permanent anti-fog and scratch-resistant coatings
  • Provides 100% UV protection and metal-free construction
  • LTX® Lens Technology for premium optical clarity
  • Progressive +2.0 diopter for magnification