Learn More
MSC HexArmor Safety Glasses, VuForce, Clear, Anti-scratch, 1 ea
Catalog No. 5000767741
Encompass
VůForce® anti-scratch safety glasses feature a full frame and built-in side shields, providing clear vision and protection in harsh conditions. These glasses offer 100% UV protection, are metal detectable, and meet stringent safety standards for various industrial applications.
  • Full frame design for a traditional look and feel
  • Side shield coverage temple arms
  • Provides 100% UV protection
  • Metal detectable
  • Vend-packed
  • Compliant with ANSI Z87.1.2020, High Impact Rated Z87+, CE EN166, and CSA Z94.3 standards
Catalog No. 50-007-67741 Supplier MSC Supplier No. 42879247
VůForce® anti-scratch safety glasses feature a full frame and built-in side shields, providing clear vision and protection in harsh conditions. These glasses offer 100% UV protection, are metal detectable, and meet stringent safety standards for various industrial applications.
  • Full frame design for a traditional look and feel
  • Side shield coverage temple arms
  • Provides 100% UV protection
  • Metal detectable
  • Vend-packed
  • Compliant with ANSI Z87.1.2020, High Impact Rated Z87+, CE EN166, and CSA Z94.3 standards