Learn More
MSC HexArmor Safety Goggles, LT300 OTG, Clear TruShield Lens, 1 ea
Catalog No. 5000789767
Encompass
The LT300 OTG Safety Goggles offer robust protection and optical clarity, designed to fit over most prescription glasses. They effectively safeguard against liquids, dirt, and debris in demanding environments.
  • Durable, complete coverage soft seal
  • Indirect channel ventilation system
  • Fits over most prescription glasses
  • Panoramic lens for wide field of vision
  • Combines soft and hard components adapting to wearer's face
  • Wide, fully adjustable head strap for pressure-free comfort
  • Compatible with other PPE, such as respirators
  • LTX® Lens Technology for premium optical clarity
  • Provides 100% UV protection
  • Completely metal-free design
  • Dual-action permanent anti-fog and scratch-resistant coatings
  • Coatings will not wash off over time
Catalog No. 50-007-89767 Supplier MSC Supplier No. 32169591
The LT300 OTG Safety Goggles offer robust protection and optical clarity, designed to fit over most prescription glasses. They effectively safeguard against liquids, dirt, and debris in demanding environments.
  • Durable, complete coverage soft seal
  • Indirect channel ventilation system
  • Fits over most prescription glasses
  • Panoramic lens for wide field of vision
  • Combines soft and hard components adapting to wearer's face
  • Wide, fully adjustable head strap for pressure-free comfort
  • Compatible with other PPE, such as respirators
  • LTX® Lens Technology for premium optical clarity
  • Provides 100% UV protection
  • Completely metal-free design
  • Dual-action permanent anti-fog and scratch-resistant coatings
  • Coatings will not wash off over time