Learn More
MSC HexArmor Safety Helmet, Class E, Short Brim, 6-Point, 1 ea
Catalog No. 5000782347
Encompass
The Ceros XP250E short brim safety helmet is a Class E helmet featuring a pre-assembled six-point suspension system. Constructed from premium HDPE, it is fully enclosed and designed for electrical work, offering advanced head protection and user comfort.
  • Type I, Class E safety helmet
  • Fully enclosed shell for electrical work
  • Short brim
  • Premium HDPE material construction
  • Pre-assembled, six-point AirBridge suspension system
  • SwiftLock one-hand-turn wheel ratchet
  • Click-and-Go magnetic and mechanical accessory integration
  • Removable, moisture-resistant, synthetic leather sweatband
  • Meets ANSI Z89.1 and CE EN397 standards
  • Class E proof tested at 20,000 volts
  • Extra back-of-head coverage
  • Includes 360-degree padded headband
Catalog No. 50-007-82347 Supplier MSC Supplier No. 13005863
The Ceros XP250E short brim safety helmet is a Class E helmet featuring a pre-assembled six-point suspension system. Constructed from premium HDPE, it is fully enclosed and designed for electrical work, offering advanced head protection and user comfort.
  • Type I, Class E safety helmet
  • Fully enclosed shell for electrical work
  • Short brim
  • Premium HDPE material construction
  • Pre-assembled, six-point AirBridge suspension system
  • SwiftLock one-hand-turn wheel ratchet
  • Click-and-Go magnetic and mechanical accessory integration
  • Removable, moisture-resistant, synthetic leather sweatband
  • Meets ANSI Z89.1 and CE EN397 standards
  • Class E proof tested at 20,000 volts
  • Extra back-of-head coverage
  • Includes 360-degree padded headband