The HexArmor Helix® 1092 tear-away glove is designed for tasks using handheld screw-driving tools, featuring Safe Finger Release® (SFR) technology with breakaway fingers. It provides protection, comfort, and full tool control, utilizing a 15-gauge nylon and elastane blend shell with a flexible foam nitrile palm coating.
- Safe Finger Release® (SFR) technology with breakaway fingers
- High-flex 15-gauge nylon and elastane blend shell provides 360° cut resistance
- Flexible foam nitrile palm coating for superior grip and abrasion resistance
- Silicone-free, REACH compliant, and safe for sensitive skin
- Elastic knit wrist helps prevent dirt and debris
- Ultimate dexterity, tactility, comfort, and breathability with seamless construction
- Launderable for extended use; ANSI/ISEA 105-2024 compliant
- Field-proven powerful grip in dry and oily situations