Learn More
MSC HexArmor VuFlight Safety Glasses, Clear, Bifocal 2.5, Anti-scratch, 1 ea
Catalog No. 5000768751
Encompass
VuFlight safety glasses are designed for reliability and comfort, featuring a wraparound lens and bifocal magnification. This specific model provides a clear lens with 2.5 magnification and an anti-scratch coating, suitable for precision tasks requiring enhanced vision.
  • Wraparound lens design
  • Flexible nose piece for comfort and non-slip fit
  • Anti-slip temple arms
  • Provides 100% UV protection
  • Anti-scratch coating
  • Compliant with ANSI Z87.1.2020, CE EN166, and CSA Z94.3
  • Metal detectable
  • Vend-packed
Catalog No. 50-007-68751 Supplier MSC Supplier No. 50732684
VuFlight safety glasses are designed for reliability and comfort, featuring a wraparound lens and bifocal magnification. This specific model provides a clear lens with 2.5 magnification and an anti-scratch coating, suitable for precision tasks requiring enhanced vision.
  • Wraparound lens design
  • Flexible nose piece for comfort and non-slip fit
  • Anti-slip temple arms
  • Provides 100% UV protection
  • Anti-scratch coating
  • Compliant with ANSI Z87.1.2020, CE EN166, and CSA Z94.3
  • Metal detectable
  • Vend-packed