Learn More
MSC HexArmor Welding Glove, Leather, 3X-Large, Stick Welding, 1 Pair
Catalog No. 5000685768
Encompass
The HeatArmor® 5051 welding glove is engineered for stick welding operations, offering 360° level A3 cut protection through its proprietary HeatHide® technology. It provides exceptional comfort, dexterity, and extended longevity for demanding applications.
  • Designed for stick welding applications.
  • Provides 360° level A3 interior cut protection.
  • HeatHide® leather treatment for extreme heat.
  • Premium heat-resistant split cowhide leather.
  • Level 2 contact heat resistance.
  • Flame-resistant materials.
  • Features 5.5 in extended gauntlet cuff with polyester lining.
  • Aramid stitching for strength and durability.
  • Good grip in dry or light oil conditions.
  • Complies with ANSI/ISEA 105-2024.
  • Maintains flexibility; resists brittleness and stiffness.
  • High dexterity and immediate comfort, no break-in required.
Catalog No. 50-006-85768 Supplier MSC Supplier No. 80804099
The HeatArmor® 5051 welding glove is engineered for stick welding operations, offering 360° level A3 cut protection through its proprietary HeatHide® technology. It provides exceptional comfort, dexterity, and extended longevity for demanding applications.
  • Designed for stick welding applications.
  • Provides 360° level A3 interior cut protection.
  • HeatHide® leather treatment for extreme heat.
  • Premium heat-resistant split cowhide leather.
  • Level 2 contact heat resistance.
  • Flame-resistant materials.
  • Features 5.5 in extended gauntlet cuff with polyester lining.
  • Aramid stitching for strength and durability.
  • Good grip in dry or light oil conditions.
  • Complies with ANSI/ISEA 105-2024.
  • Maintains flexibility; resists brittleness and stiffness.
  • High dexterity and immediate comfort, no break-in required.