Learn More
Medchemexpress LLC Hexatriacontane | 630-06-8 | MFCD00009413 | 98.0% | 506.97 g/mol | C36H74 | 25 G
SDP
Catalog No. 5000283113
Encompass_Preferred
Hexatriacontane (C36 alkane) is a long-chain saturated hydrocarbon used as a reagent and analytical standard in research and quality control. It is supplied as a solid with documented purity and characterization information for laboratory handling and analysis.
  • High purity (98.0%) reagent suitable for analytical and preparative applications.
  • Long-chain saturated hydrocarbon; chemical formula C36H74; molecular weight 506.97 g/mol.
  • Provided as a solid 25 G quantity for laboratory use.
  • CAS number 630-06-8 for unambiguous substance identification.
  • Documentation available: data sheet, certificate of analysis, safety data sheet, and HNMR on request.
Catalog No. 50-002-83113 Supplier Medchemexpress LLC Supplier No. HYW13135325G
Hexatriacontane (C36 alkane) is a long-chain saturated hydrocarbon used as a reagent and analytical standard in research and quality control. It is supplied as a solid with documented purity and characterization information for laboratory handling and analysis.
  • High purity (98.0%) reagent suitable for analytical and preparative applications.
  • Long-chain saturated hydrocarbon; chemical formula C36H74; molecular weight 506.97 g/mol.
  • Provided as a solid 25 G quantity for laboratory use.
  • CAS number 630-06-8 for unambiguous substance identification.
  • Documentation available: data sheet, certificate of analysis, safety data sheet, and HNMR on request.