Learn More
Medchemexpress LLC Hexatriacontane-d74 | 16416-34-5 | MFCD00274270 | 99.0% | 581.43 g/mol | C36D74 | 100 MG
SDP
Catalog No. 5000283267
Encompass_Preferred
Hexatriacontane-d74 is a deuterium-labeled C36 alkane supplied as a high-purity stable isotope standard for analytical and tracer applications. It is intended for use as an internal standard or tracer in quantitative NMR, GC-MS, and LC-MS analyses, and for studies where isotope labeling aids pharmacokinetic or metabolic investigation. Documentation such as COA, SDS, and data sheets is available from the supplier.
Catalog No. 50-002-83267 Supplier Medchemexpress LLC Supplier No. HYW131353S100MG
Hexatriacontane-d74 is a deuterium-labeled C36 alkane supplied as a high-purity stable isotope standard for analytical and tracer applications. It is intended for use as an internal standard or tracer in quantitative NMR, GC-MS, and LC-MS analyses, and for studies where isotope labeling aids pharmacokinetic or metabolic investigation. Documentation such as COA, SDS, and data sheets is available from the supplier.