Learn More
Medchemexpress LLC Hexatriacontane-d74 | 16416-34-5 | MFCD00274270 | 99.0% | 581.43 g/mol | CD3(CD2)34CD3 | 50 MG
SDP
Catalog No. 5000283300
Encompass_Preferred
Hexatriacontane-d74 is the deuterium-labeled isotopologue of hexatriacontane (C36) supplied as a high-purity solid for use as an isotopic standard or tracer in analytical chemistry. It is designed for quantitative and qualitative mass-spectrometry applications where a clear mass shift and stable reference compound are required.
  • High isotopic purity (99.0%).
  • Deuterium-labeled C36 alkane for clear mass-shift separation.
  • Solid form for stable storage and handling.
  • Suitable as an internal standard in GC-MS and LC-MS workflows.
  • Provided with certificate of analysis and safety data sheet for documentation.
  • Useful for tracer studies and analytical method development.
Catalog No. 50-002-83300 Supplier Medchemexpress LLC Supplier No. HYW131353S50MG
Hexatriacontane-d74 is the deuterium-labeled isotopologue of hexatriacontane (C36) supplied as a high-purity solid for use as an isotopic standard or tracer in analytical chemistry. It is designed for quantitative and qualitative mass-spectrometry applications where a clear mass shift and stable reference compound are required.
  • High isotopic purity (99.0%).
  • Deuterium-labeled C36 alkane for clear mass-shift separation.
  • Solid form for stable storage and handling.
  • Suitable as an internal standard in GC-MS and LC-MS workflows.
  • Provided with certificate of analysis and safety data sheet for documentation.
  • Useful for tracer studies and analytical method development.