Learn More
Bachem H-Tyr-Tyr-Tyr-Tyr-Tyr-Tyr-OH
SDP
Supplier:  Bachem 4035437.0005
Encompass_Preferred
H-Tyr-Tyr-Tyr-Tyr-Tyr-Tyr-OH Trifluoroacetate, 6934-38-9, C54H56N6O13, Mr 997.06, 5mg. Hexa-L-tyrosine. Hexatyrosine has been used as a model peptide, e.g., in mass spectrometry.
Catalog No. 50-194-7521