Learn More
ERA WATERS CORP HEXAVALENT CHROMIUM PT SAMPLE
SDP
Supplier:  ERA WATERS CORP 984
Encompass
One 15 mL screw-cap vial yields up to 2 liters after dilution. Use with IC or colorimetric methods.
Catalog No. NC9587534