Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexavalent Chromium, standard solution, Specpure™ Cr∣+6 1000μg/mL

Catalog No. AA42234AE
Click to view available options
Quantity:
100 mL
500 mL

Catalog No. AA42234AE

Supplier: Thermo Scientific Chemicals 042234AE

Only null left
Add to Cart

Hexavalent Chromium, standard solution is used in ion chromatography for measurement and determination of soluble Cr(VI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00717544

Specifications

Odor Odorless
Physical Form Liquid
Color Yellow
Solubility Miscible with water
Quantity 100 mL
Grade Specpure
Chemical Name or Material Hexavalent Chromium standard solution, Cr+6 1000μg/mL
Hazard Category H340-H350-H360
Hazard Statement GHS H Statement
H334-H340-H350-H360-H317-H208
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 232-143-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.