Learn More
Hach Company CDTA Digital Titrator Cartridge, 0.0800 M
Supplier:  Hach Company 1440201
Encompass
Cyclohexanediamine tetraacetic acid standard solution. For Calcium and Total Hardness determination by Digital Titrator titration. 0.0800 M. Digits x 0.1 = mg/L CaCO3 in 100 mL. Digits x 0.0001 = mmole.
Catalog No. 50-199-5026