Learn More
Hach Company CDTA Digital Titrator Cartridge, 0.800 M
Supplier: Hach Company 1440301
Cyclohexanediamine tetraacetic acid standard solution. For Calcium and Total Hardness determination by Digital Titrator titration. 0.800 M. Digits = mg/L CaCO3 in 100 mL. Digits x 0.001 = mmole.