Learn More
Medchemexpress LLC Hexazinone-d6 | 1219804-22-4 | 98.9% | 258.35 | C12H14D6N4O2 | 10 MG
SDP
Catalog No. 5000298904
Encompass_Preferred
Hexazinone-d6 is the deuterium-labeled analogue of hexazinone supplied as an analytical standard for mass spectrometry, quantification, and tracer studies. It is provided as a small neat solid with high isotopic enrichment suitable for laboratory analytical workflows.
  • Deuterium-labeled (d6) internal standard for mass spectrometry analyses.
  • High purity and isotopic labeling suitable for quantitative assays.
  • Intended for analytical and research use, including tracer and quantification studies.
  • Provided in a small neat package for convenient laboratory use.
Catalog No. 50-002-98904 Supplier Medchemexpress LLC Supplier No. HYB1849S10MG
Hexazinone-d6 is the deuterium-labeled analogue of hexazinone supplied as an analytical standard for mass spectrometry, quantification, and tracer studies. It is provided as a small neat solid with high isotopic enrichment suitable for laboratory analytical workflows.
  • Deuterium-labeled (d6) internal standard for mass spectrometry analyses.
  • High purity and isotopic labeling suitable for quantitative assays.
  • Intended for analytical and research use, including tracer and quantification studies.
  • Provided in a small neat package for convenient laboratory use.