Learn More
Medchemexpress LLC Hexazinone-d6 | 1219804-22-4 | 98.9% | 5 MG
SDP
Catalog No. 5000409780
Encompass_Preferred
Hexazinone-d6 is the deuterium labeled Hexazinone, a nonselective herbicide from the triazine family. It inhibits photosynthesis by binding to the D-1 quinone protein of the electron transport chain in photosystem II.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-09780 Supplier Medchemexpress LLC Supplier No. HYB1849S5MG
Hexazinone-d6 is the deuterium labeled Hexazinone, a nonselective herbicide from the triazine family. It inhibits photosynthesis by binding to the D-1 quinone protein of the electron transport chain in photosystem II.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS