Learn More
Aviva Systems Biology Corp HEXD Antibody - middle region (ARP55666_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55666_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HEXDC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HEXDC. Alternate names: FLJ23825ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61571