Learn More
Apexbio Technology LLC Hexestrol 84-16-2 200mg
SDP
Catalog No. 5000087658
Encompass_Preferred
Hexestrol (CAS 84-16-2) is a small-molecule agonist targeting estrogen receptors ER and ER It is designed to selectively activate these receptors thereby regulating hormone-responsive gene expression and associated biological pathways Hexestrol exerts its biological activity primarily through direct agonist-mediated activation of ER and ER In in vitro studies Hexestrol demonstrates potent activation with reported EC50 values of approximately 0 07 nM for ER and 0 175 nM for ER Based on these pharmacological properties Hexestrol holds research potential in studies involving estrogen receptor signaling hormone-dependent cellular responses and pathologies related to estrogen deficiency
Catalog No. 50-000-87658 Supplier Apexbio Technology LLC Supplier No. B1509200
Hexestrol (CAS 84-16-2) is a small-molecule agonist targeting estrogen receptors ER and ER It is designed to selectively activate these receptors thereby regulating hormone-responsive gene expression and associated biological pathways Hexestrol exerts its biological activity primarily through direct agonist-mediated activation of ER and ER In in vitro studies Hexestrol demonstrates potent activation with reported EC50 values of approximately 0 07 nM for ER and 0 175 nM for ER Based on these pharmacological properties Hexestrol holds research potential in studies involving estrogen receptor signaling hormone-dependent cellular responses and pathologies related to estrogen deficiency