Learn More
Medchemexpress LLC Hexestrol | 84-16-2 | 99.79% | 50 MG
SDP
Catalog No. 5000413145
Encompass_Preferred
Hexestrol is a nonsteroidal synthetic estrogen with a Ki of 0.06 and 0.06 nM for estrogen receptor alpha (ERα) and ERβ. It can be used for the research of diseases caused by estrogen deficiency and can also increase the weight of cattle.
  • Nonsteroidal synthetic estrogen
  • Ki of 0.06 and 0.06 nM for estrogen receptor alpha (ERα) and ERβ
  • Can be used for research of diseases caused by estrogen deficiency
  • Can increase the weight of cattle
Catalog No. 50-004-13145 Supplier Medchemexpress LLC Supplier No. HYB166250MG
Hexestrol is a nonsteroidal synthetic estrogen with a Ki of 0.06 and 0.06 nM for estrogen receptor alpha (ERα) and ERβ. It can be used for the research of diseases caused by estrogen deficiency and can also increase the weight of cattle.
  • Nonsteroidal synthetic estrogen
  • Ki of 0.06 and 0.06 nM for estrogen receptor alpha (ERα) and ERβ
  • Can be used for research of diseases caused by estrogen deficiency
  • Can increase the weight of cattle