Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexestrol 98.0+%, TCI America™
SDP

Catalog No. D049410G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D049410G 10 g
D04941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D049410G Supplier TCI America Supplier No. D049410G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84-16-2
Molecular Formula C18H22O2
Molecular Weight (g/mol) 270.37
MDL Number MFCD00068996
InChI Key PBBGSZCBWVPOOL-UHFFFAOYNA-N
Synonym hexestrol, meso-hexestrol, hexoestrol, hexanoestrol, hexestrofen, mesohexestrol, erythrohexestrol, estra-plex, cycloestrol, estrifar
PubChem CID 192197
IUPAC Name 4-[4-(4-hydroxyphenyl)hexan-3-yl]phenol
SMILES CCC(C(CC)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 10 g
Formula Weight 270.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexestrol
RTECSNumber SL0560850
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.