Learn More
Aat Bioquest Hexidium Iodide CAS 211566-66-4 (5 mg)
SDP
Catalog No. 502249749
Encompass_Preferred
The nucleic acid stain hexidium iodide is permeant to mammalian cells and selectively stains almost all gram-positive bacteria in the presence of gram-negative bacteria. This catalog is for 5 mg.
This item is not returnable. View return policy
Catalog No. 50-224-9749 Supplier Aat Bioquest Supplier No. 17562
The nucleic acid stain hexidium iodide is permeant to mammalian cells and selectively stains almost all gram-positive bacteria in the presence of gram-negative bacteria. This catalog is for 5 mg.