Learn More
Medchemexpress LLC Hexidium iodide | 1MG
SDP
Catalog No. 5000226679
Encompass_Preferred
Hexidium iodide a fluorescent nucleic binding acid stain (excitation/emission 518/600 nm) permeants to mammalian cells and selectively stains almost all gram-positive bacteria Hexidium iodide can bind to the DNA of all bacteria after permeabilization by EDTA[1][2]
Catalog No. 50-002-26679 Supplier Medchemexpress LLC Supplier No. HY1142271MG
Hexidium iodide a fluorescent nucleic binding acid stain (excitation/emission 518/600 nm) permeants to mammalian cells and selectively stains almost all gram-positive bacteria Hexidium iodide can bind to the DNA of all bacteria after permeabilization by EDTA[1][2]