Learn More
ABclonal Technology HEXIM1 Rabbit mAb
SDP
Catalog No. 5000005494
Encompass_Preferred
Expression of this gene is induced by hexamethylene-bis-acetamide in vascular smooth muscle cells This gene has no introns
Catalog No. 50-000-05494 Supplier ABclonal Technology Supplier No. A2420820UL
Expression of this gene is induced by hexamethylene-bis-acetamide in vascular smooth muscle cells This gene has no introns