Learn More
Cayman Chemical HexnoIc AcId-d3 10mg
SDP
Catalog No. 502937713
Encompass_Preferred
An internal standard for the quantification of hexanoic acid by GC- or LC-MS
Catalog No. 50-293-7713 Supplier Cayman Chemical Supplier No. 2849710 MG
An internal standard for the quantification of hexanoic acid by GC- or LC-MS