Learn More
Cayman Chemical Hexanoic Acid-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 55320-69-9, 50 mg
SDP
Catalog No. 502937804
Encompass_Preferred
An internal standard for the quantification of hexanoic acid by GC- or LC-MS
Catalog No. 50-293-7804 Supplier Cayman Chemical Supplier No. 2849750 MG
An internal standard for the quantification of hexanoic acid by GC- or LC-MS