Learn More
Cayman Chemical HexnoIc AcId-d5 50mg
SDP
Catalog No. 502991960
Encompass_Preferred
An internal standard for the quantification of hexanoicacid by GC- or LC-MS
Catalog No. 50-299-1960 Supplier Cayman Chemical Supplier No. 3124850 MG
An internal standard for the quantification of hexanoicacid by GC- or LC-MS