Learn More
Cayman Chemical HexnolamIno PAF C-16 10mg
SDP
Catalog No. 502893718
Encompass_Preferred
Hexanolamino PAF C-16 is a PAF analog with mixed PAF receptor agonist/antagonist properties. In human monocyte-derived macrophages, it is an antagonist which inhibits the production of reactive oxygen species in response to PAF C-16.{367} On the other hand, in rabbit platelets and guinea pig macrophages, hexanolamino PAF C-16 is a partial agonist, and in guinea pig platelets it is a full agonist.{3786}
Catalog No. 50-289-3718 Supplier Cayman Chemical Supplier No. 6090510 MG
Hexanolamino PAF C-16 is a PAF analog with mixed PAF receptor agonist/antagonist properties. In human monocyte-derived macrophages, it is an antagonist which inhibits the production of reactive oxygen species in response to PAF C-16.{367} On the other hand, in rabbit platelets and guinea pig macrophages, hexanolamino PAF C-16 is a partial agonist, and in guinea pig platelets it is a full agonist.{3786}