Learn More
Cayman Chemical Hexnoyl-Coenzyme AsodIum s 1mg
SDP
Catalog No. 502940980
Encompass_Preferred
A medium-chain fatty acyl CoA; the preferred acyl donor substrate for GOAT; a precursor in the biosynthesis of olivetolic acid
Catalog No. 50-294-0980 Supplier Cayman Chemical Supplier No. 278661 MG
A medium-chain fatty acyl CoA; the preferred acyl donor substrate for GOAT; a precursor in the biosynthesis of olivetolic acid