Learn More
Cayman Chemical Hexnoyl-Coenzyme AsodIum s 5mg
SDP
Catalog No. 502939212
Encompass_Preferred
A medium-chain fatty acyl CoA; the preferred acyl donor substrate for GOAT; a precursor in the biosynthesis of olivetolic acid
Catalog No. 50-293-9212 Supplier Cayman Chemical Supplier No. 278665 MG
A medium-chain fatty acyl CoA; the preferred acyl donor substrate for GOAT; a precursor in the biosynthesis of olivetolic acid