Learn More
Cayman Chemical Hexanoyl Glycine, Purity: ≥95%, CAS: 24003-67-6, 10 mg
SDP
Catalog No. 502969889
Encompass_Preferred
An acylated amino acid that is used as a urinary biomarker for several indications; increased urinary excretion is indicative of MCAD deficiency; can also be used as a biomarker for exposure to gamma radiation
Catalog No. 50-296-9889 Supplier Cayman Chemical Supplier No. 1641210 MG
An acylated amino acid that is used as a urinary biomarker for several indications; increased urinary excretion is indicative of MCAD deficiency; can also be used as a biomarker for exposure to gamma radiation