Learn More
Cayman Chemical Hexnoyl-L-carnItInchlorId 10mg
SDP
Catalog No. 502933480
Encompass_Preferred
A medium-chain acylcarnitine; has been used in the synthesis of acylcarnitine benzyl esters
Catalog No. 50-293-3480 Supplier Cayman Chemical Supplier No. 2655410 MG
A medium-chain acylcarnitine; has been used in the synthesis of acylcarnitine benzyl esters