Learn More
Cayman Chemical Hexanoyl-L-carnitine (chloride), Purity: ≥95%, CAS: 162067-53-0, 50 mg
SDP
Catalog No. 502935463
Encompass_Preferred
A medium-chain acylcarnitine; has been used in the synthesis of acylcarnitine benzyl esters
Catalog No. 50-293-5463 Supplier Cayman Chemical Supplier No. 2655450 MG
A medium-chain acylcarnitine; has been used in the synthesis of acylcarnitine benzyl esters