Learn More
Cayman Chemical Hexanoyl-L-carnitine (chloride), Purity: ≥95%, CAS: 162067-53-0, 5 mg
SDP
Catalog No. 502934545
Encompass_Preferred
A medium-chain acylcarnitine; has been used in the synthesis of acylcarnitine benzyl esters
Catalog No. 50-293-4545 Supplier Cayman Chemical Supplier No. 265545 MG
A medium-chain acylcarnitine; has been used in the synthesis of acylcarnitine benzyl esters