Learn More
Cayman Chemical HexnoylCoenzyme AsodIum s 10mg
SDP
Catalog No. 502939111
Encompass_Preferred
A medium-chain fatty acyl CoA; the preferred acyl donor substrate for GOAT; a precursor in the biosynthesis of olivetolic acid
Catalog No. 50-293-9111 Supplier Cayman Chemical Supplier No. 2786610 MG
A medium-chain fatty acyl CoA; the preferred acyl donor substrate for GOAT; a precursor in the biosynthesis of olivetolic acid