Learn More
Sigma Aldrich Fine Chemicals Biosciences Hexokinase/Glucose-6-Phosp
Catalog No. 502905345
Encompass
Hexokinase is responsible for catalyzing the first step of glycolysis. It results in the formation of glucose-6-phosphate and ADP from glucose and ATP. Glucose-6-phosphate dehydrogenase is a crucial enzyme in the pentose phosphate pathway. It is responsible for the conversion of glucose-6-phosphate and NADP (nicotinamide adenine dinucleotide phosphate) to 6-phosphogluconolactone and NADPH.
Catalog No. 50-290-5345 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10127825001
Hexokinase is responsible for catalyzing the first step of glycolysis. It results in the formation of glucose-6-phosphate and ADP from glucose and ATP. Glucose-6-phosphate dehydrogenase is a crucial enzyme in the pentose phosphate pathway. It is responsible for the conversion of glucose-6-phosphate and NADP (nicotinamide adenine dinucleotide phosphate) to 6-phosphogluconolactone and NADPH.