Learn More
Medchemexpress LLC Hexokinase I Antibody | Affinity purified | 102 kDa | 10 UL
SDP
Catalog No. 5000395856
Encompass_Preferred
Hexokinase I Antibody (YA739) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting Hexokinase I. Hexokinases phosphorylate glucose to produce glucose-6-phosphate, the first step in most glucose metabolism pathways. This gene encodes a ubiquitous form of hexokinase which localizes to the outer membrane of mitochondria.
  • Mouse-derived and non-conjugated IgG1 monoclonal antibody
  • Targets Hexokinase I
  • Reacts with human, mouse, and rat samples
  • Localizes to the outer membrane of mitochondria
  • Associated with hemolytic anemia due to hexokinase deficiency when mutated
  • Supplied in PBS, pH 7.4
  • Stored at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Application: WB with a dilution ratio of 1:500-1:1000
Catalog No. 50-003-95856 Supplier Medchemexpress LLC Supplier No. HYP8070210UL
Hexokinase I Antibody (YA739) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting Hexokinase I. Hexokinases phosphorylate glucose to produce glucose-6-phosphate, the first step in most glucose metabolism pathways. This gene encodes a ubiquitous form of hexokinase which localizes to the outer membrane of mitochondria.
  • Mouse-derived and non-conjugated IgG1 monoclonal antibody
  • Targets Hexokinase I
  • Reacts with human, mouse, and rat samples
  • Localizes to the outer membrane of mitochondria
  • Associated with hemolytic anemia due to hexokinase deficiency when mutated
  • Supplied in PBS, pH 7.4
  • Stored at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Application: WB with a dilution ratio of 1:500-1:1000