Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexokinase II Antibody (YA386) | 102 kDa | 10 UL
SDP

Catalog No. 5000395799
Encompass_Preferred

Hexokinase II Antibody (YA386) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, specifically targeting Hexokinase II. This antibody is instrumental in studying cellular metabolism, as Hexokinase II catalyzes the initial phosphorylation of hexoses like D-glucose and D-fructose to their 6-phosphate forms, a critical step in glycolysis. It also contributes to maintaining the structural integrity of the outer mitochondrial membrane.

  • Rabbit-derived monoclonal IgG antibody
  • Targets Hexokinase II
  • Affinity purified
  • Molecular weight: 102 kDa
  • Reacts with Human, Rat, Hamster
  • Applications: WB (1:500-1:1000), IHC-P (1:50-1:100), IP (1:20)
  • Formulation: 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.05% BSA, 0.01% Sodium azide
  • Storage: -20°C for 1 year, avoid freeze/thaw cycles

Catalog No. 50-003-95799 Supplier Medchemexpress LLC Supplier No. HYP8070310UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexokinase II Antibody (YA386) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, specifically targeting Hexokinase II. This antibody is instrumental in studying cellular metabolism, as Hexokinase II catalyzes the initial phosphorylation of hexoses like D-glucose and D-fructose to their 6-phosphate forms, a critical step in glycolysis. It also contributes to maintaining the structural integrity of the outer mitochondrial membrane.

  • Rabbit-derived monoclonal IgG antibody
  • Targets Hexokinase II
  • Affinity purified
  • Molecular weight: 102 kDa
  • Reacts with Human, Rat, Hamster
  • Applications: WB (1:500-1:1000), IHC-P (1:50-1:100), IP (1:20)
  • Formulation: 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.05% BSA, 0.01% Sodium azide
  • Storage: -20°C for 1 year, avoid freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.