Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexokinase II Antibody (YA386) | 102 kDa | 100 UL
SDP

Catalog No. 5000395557
Encompass_Preferred

This non-conjugated IgG monoclonal antibody, derived from rabbits, specifically targets Hexokinase II. It is intended for research use only. The antibody is supplied in a buffered solution containing Tris-Glycine, NaCl, Glycerol, BSA, and sodium azide as a preservative, ensuring stability and performance. It is reactive with human, rat, and hamster species, and its immunogen is a synthetic peptide corresponding to human Hexokinase II.

  • Reactive with human, rat, and hamster species
  • Immunogen is a synthetic peptide corresponding to human Hexokinase II
  • Applications include Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), and Immunoprecipitation (IP)
  • Stored at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Shipped with blue ice

Catalog No. 50-003-95557 Supplier Medchemexpress LLC Supplier No. HYP80703100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This non-conjugated IgG monoclonal antibody, derived from rabbits, specifically targets Hexokinase II. It is intended for research use only. The antibody is supplied in a buffered solution containing Tris-Glycine, NaCl, Glycerol, BSA, and sodium azide as a preservative, ensuring stability and performance. It is reactive with human, rat, and hamster species, and its immunogen is a synthetic peptide corresponding to human Hexokinase II.

  • Reactive with human, rat, and hamster species
  • Immunogen is a synthetic peptide corresponding to human Hexokinase II
  • Applications include Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), and Immunoprecipitation (IP)
  • Stored at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Shipped with blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.