Learn More
Medchemexpress LLC Hexokinase II Antibody (YA4082) | 102 kDa | 10 UL
SDP
Catalog No. 5000446130
Encompass_Preferred
Hexokinase II Antibody (YA4082) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting Hexokinase II. It is intended for research use only. Hexokinase II (HK2) catalyzes the phosphorylation of hexose, such as D-glucose and D-fructose, to hexose 6-phosphate. It mediates the initial step of glycolysis by catalyzing the phosphorylation of D-glucose to D-glucose 6-phosphate.
  • Reacts with Human, Mouse, Rat, and Rabbit species
  • Predicted and observed band size is 102 kDa
  • Supplied as a liquid in PBS with 0.05% sodium azide
  • Applications include WB, IHC-P, FC, and ELISA
  • Key role in maintaining the integrity of the outer mitochondrial membrane
  • Subcellular localization includes the mitochondrion outer membrane and the cytoplasm
Catalog No. 50-004-46130 Supplier Medchemexpress LLC Supplier No. HYP8438510UL
Hexokinase II Antibody (YA4082) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting Hexokinase II. It is intended for research use only. Hexokinase II (HK2) catalyzes the phosphorylation of hexose, such as D-glucose and D-fructose, to hexose 6-phosphate. It mediates the initial step of glycolysis by catalyzing the phosphorylation of D-glucose to D-glucose 6-phosphate.
  • Reacts with Human, Mouse, Rat, and Rabbit species
  • Predicted and observed band size is 102 kDa
  • Supplied as a liquid in PBS with 0.05% sodium azide
  • Applications include WB, IHC-P, FC, and ELISA
  • Key role in maintaining the integrity of the outer mitochondrial membrane
  • Subcellular localization includes the mitochondrion outer membrane and the cytoplasm