Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexokinase II antibody (YA4082) | 102 kDa | 50 UL
SDP

Catalog No. 5000447609
Encompass_Preferred

Hexokinase II antibody (YA4082) is a mouse-derived, non-conjugated IgG1 monoclonal antibody. Designed for research use, it targets Hexokinase II, an enzyme essential for glycolysis and maintaining outer mitochondrial membrane integrity. This antibody is supplied in PBS with 0.05% sodium azide and is compatible with Western Blot, IHC-P, Flow Cytometry, and ELISA applications.

  • Mouse-derived IgG1 monoclonal antibody
  • Targets Hexokinase II
  • Reactive with human, mouse, rat, rabbit species
  • Catalyzes hexose phosphorylation, initiating glycolysis
  • Maintains outer mitochondrial membrane integrity
  • Applications: WB, IHC-P, FC, and ELISA
  • Storage: -20°C for 1 year
  • Affinity purified

Catalog No. 50-004-47609 Supplier Medchemexpress LLC Supplier No. HYP8438550UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexokinase II antibody (YA4082) is a mouse-derived, non-conjugated IgG1 monoclonal antibody. Designed for research use, it targets Hexokinase II, an enzyme essential for glycolysis and maintaining outer mitochondrial membrane integrity. This antibody is supplied in PBS with 0.05% sodium azide and is compatible with Western Blot, IHC-P, Flow Cytometry, and ELISA applications.

  • Mouse-derived IgG1 monoclonal antibody
  • Targets Hexokinase II
  • Reactive with human, mouse, rat, rabbit species
  • Catalyzes hexose phosphorylation, initiating glycolysis
  • Maintains outer mitochondrial membrane integrity
  • Applications: WB, IHC-P, FC, and ELISA
  • Storage: -20°C for 1 year
  • Affinity purified

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.