Learn More
Medchemexpress LLC Hexokinase II VDAC binding domain peptide | 99.9% | 3997.78 | 1 MG
SDP
Catalog No. 5000423432
Encompass_Preferred
Hexokinase II VDAC binding domain peptide (Hxk2VBD peptide) is a cell-permeable hexokinase II VDAC binding domain. It inhibits mitochondrial localization of hexokinase 2 (HXK2) and neurotrophic factor-directed axon outgrowth.
  • Cell-permeable
  • Inhibits mitochondrial localization of hexokinase 2 (HXK2)
  • Inhibits neurotrophic factor-directed axon outgrowth
  • White to off-white solid appearance
Catalog No. 50-004-23432 Supplier Medchemexpress LLC Supplier No. HYP101081MG
Hexokinase II VDAC binding domain peptide (Hxk2VBD peptide) is a cell-permeable hexokinase II VDAC binding domain. It inhibits mitochondrial localization of hexokinase 2 (HXK2) and neurotrophic factor-directed axon outgrowth.
  • Cell-permeable
  • Inhibits mitochondrial localization of hexokinase 2 (HXK2)
  • Inhibits neurotrophic factor-directed axon outgrowth
  • White to off-white solid appearance