Learn More
Medchemexpress LLC Hexokinase II VDAC binding domain peptide, cell-permeable | 99.97% | 3997.78 | 5 MG
SDP
Catalog No. 5000422565
Encompass_Preferred
Hexokinase II VDAC binding domain peptide (Hxk2VBD peptide) is a cell-permeable hexokinase II VDAC binding domain. It inhibits the mitochondrial localization of hexokinase 2 (HXK2) and neurotrophic factor-directed axon outgrowth.
  • Cell-permeable hexokinase II VDAC binding domain
  • Inhibits mitochondrial localization of hexokinase 2
  • Inhibits neurotrophic factor-directed axon outgrowth
  • For research use only
Catalog No. 50-004-22565 Supplier Medchemexpress LLC Supplier No. HYP101085MG
Hexokinase II VDAC binding domain peptide (Hxk2VBD peptide) is a cell-permeable hexokinase II VDAC binding domain. It inhibits the mitochondrial localization of hexokinase 2 (HXK2) and neurotrophic factor-directed axon outgrowth.
  • Cell-permeable hexokinase II VDAC binding domain
  • Inhibits mitochondrial localization of hexokinase 2
  • Inhibits neurotrophic factor-directed axon outgrowth
  • For research use only