Learn More
Medchemexpress LLC Hexokinase, microorganism | 9001-51-8 | ≥500 U/mg protein | 1 KU
SDP
Catalog No. 5000400333
Encompass_Preferred
Hexokinase (ScHEX1) (EC 2.7.1.1) is a glycolytic enzyme that is inhibited by n-acetylglucosamine. This inhibition leads to its separation from the mitochondrial outer membrane, which results in the activation of NLRP3 inflammasome.
Catalog No. 50-004-00333 Supplier Medchemexpress LLC Supplier No. HYP27761KU
Hexokinase (ScHEX1) (EC 2.7.1.1) is a glycolytic enzyme that is inhibited by n-acetylglucosamine. This inhibition leads to its separation from the mitochondrial outer membrane, which results in the activation of NLRP3 inflammasome.