Learn More
Medchemexpress LLC Hexokinase, yeast | 9001-51-8 | 1 KU
SDP
Catalog No. 5000397375
Encompass_Preferred
Hexokinase, Yeast catalyzes the phosphorylation of glucose and is involved in glycolysis. It also participates in energy production, protection of mitochondrial integrity, and cell survival. This product is for research use only and is not sold to patients.
  • Catalyzes the phosphorylation of glucose
  • Involved in glycolysis
  • Participates in energy production
  • Protection of mitochondrial integrity
  • Supports cell survival
  • For research use only
Catalog No. 50-003-97375 Supplier Medchemexpress LLC Supplier No. HYP2776A1KU
Hexokinase, Yeast catalyzes the phosphorylation of glucose and is involved in glycolysis. It also participates in energy production, protection of mitochondrial integrity, and cell survival. This product is for research use only and is not sold to patients.
  • Catalyzes the phosphorylation of glucose
  • Involved in glycolysis
  • Participates in energy production
  • Protection of mitochondrial integrity
  • Supports cell survival
  • For research use only