Learn More
Medchemexpress LLC Hexokinase, Yeast | 9001-51-8 | 5 KU
SDP
Catalog No. 5000400400
Encompass_Preferred
Hexokinase, Yeast catalyzes the phosphorylation of glucose and is involved in glycolysis. It also contributes to energy production, protection of mitochondrial integrity, and cell survival.
  • Catalyzes phosphorylation of glucose
  • Involved in glycolysis
  • Contributes to energy production
  • Protects mitochondrial integrity
  • Supports cell survival
  • Appearance: Solid
  • Color: White to off-white
  • Intended for research use only
Catalog No. 50-004-00400 Supplier Medchemexpress LLC Supplier No. HYP2776A5KU
Hexokinase, Yeast catalyzes the phosphorylation of glucose and is involved in glycolysis. It also contributes to energy production, protection of mitochondrial integrity, and cell survival.
  • Catalyzes phosphorylation of glucose
  • Involved in glycolysis
  • Contributes to energy production
  • Protects mitochondrial integrity
  • Supports cell survival
  • Appearance: Solid
  • Color: White to off-white
  • Intended for research use only