Learn More
Medchemexpress LLC Hexokinase, Yeast | 9001-51-8 | MFCD00131321 | 10 KU
SDP
Catalog No. 5000397347
Encompass_Preferred
Hexokinase, Yeast catalyzes the phosphorylation of glucose and is involved in glycolysis. It is also related to energy production, protection of mitochondrial integrity, and cell survival.
  • Catalyzes the phosphorylation of glucose.
  • Involved in glycolysis.
  • Related to energy production.
  • Contributes to the protection of mitochondrial integrity.
  • Contributes to cell survival.
  • For research use only.
Catalog No. 50-003-97347 Supplier Medchemexpress LLC Supplier No. HYP2776A10KU
Hexokinase, Yeast catalyzes the phosphorylation of glucose and is involved in glycolysis. It is also related to energy production, protection of mitochondrial integrity, and cell survival.
  • Catalyzes the phosphorylation of glucose.
  • Involved in glycolysis.
  • Related to energy production.
  • Contributes to the protection of mitochondrial integrity.
  • Contributes to cell survival.
  • For research use only.